Assay Details
| Gene Symbol: Ptcd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027275.3 | NP_081551.2 | 14 - 15 | 1169 | 137 | |
| GenBank mRNA | AK032831.1 | - | 14 - 15 | 1153 | 137 | |
| AK146005.1 | - | 14 - 15 | 1158 | 137 | ||
| BC060687.1 | - | 5 - 6 | 390 | 137 | ||
| BC084595.1 | - | 5 - 6 | 419 | 137 | ||
More Information
Set Membership: |
|

Ptcd3
Genomic Map