Assay Details
| Gene Symbol: Ptcd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027275.3 | NP_081551.2 | 17 - 18 | 1400 | 148 | |
| GenBank mRNA | AK013131.1 | - | 3 - 4 | 179 | 148 | |
| AK146005.1 | - | 17 - 18 | 1389 | 148 | ||
| AK146172.1 | - | 17 - 18 | 148 | |||
| AK172480.1 | - | 18 - 19 | 1465 | 148 | ||
| BC055844.1 | - | 3 - 4 | 227 | 148 | ||
| BC060687.1 | - | 8 - 9 | 621 | 148 | ||
| BC084595.1 | - | 8 - 9 | 650 | 148 | ||
| BC115434.2 | - | 17 - 18 | 1396 | 40094203 | 148 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ptcd3
Genomic Map