Assay Details
| Gene Symbol: Ptcd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027275.3 | NP_081551.2 | 7 - 8 | 558 | 109 | |
| GenBank mRNA | AK032831.1 | - | 7 - 8 | 542 | 109 | |
| AK146005.1 | - | 7 - 8 | 547 | 109 | ||
| AK146172.1 | - | 7 - 8 | 559 | 109 | ||
| AK172480.1 | - | 7 - 8 | 547 | 109 | ||
| BC115434.2 | - | 7 - 8 | 554 | 40094203 | 109 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ptcd3
Genomic Map