Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0600006534.mm.1 |
TC0600000852.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600006534.mm.1 |
NM_001271395.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600006536.mm.1 |
TC0600000852.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600006536.mm.1 |
NM_001042534.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600006536.mm.1 |
NM_001271395.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600006536.mm.1 |
NM_001271415.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600006536.mm.1 |
NM_007599.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600006533.mm.1 |
TC0600000852.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600006533.mm.1 |
NM_001271395.1 |
Refseq |