Assay Details
| Gene Symbol: Parp11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181402.3 | NP_852067.1 | 7 - 8 | 747 | 150 | |
| GenBank mRNA | AK030559.1 | - | 1 - 2 | 124 | 150 | |
| AK151393.1 | - | 4 - 5 | 437 | 150 | ||
| BC040269.1 | - | 7 - 8 | 747 | 4035469 | 150 | |
More Information
Updated Mapping Information
Set Membership: |
|

Parp11
Genomic Map