Assay Details
| Gene Symbol: Ltbr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010736.3 | NP_034866.1 | 8 - 9 | 972 | 78 | |
| GenBank mRNA | AK078859.1 | - | 4 - 5 | 231 | 78 | |
| AK146127.1 | - | 8 - 9 | 955 | 78 | ||
| BC138590.1 | - | 8 - 9 | 835 | 8861610 | 78 | |
| BC138591.1 | - | 8 - 9 | 835 | 8861611 | 78 | |
| L38423.1 | - | 8 - 9 | 972 | 78 | ||
| U29173.1 | - | 8 - 9 | 956 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ltbr
Genomic Map