Assay Details
| Gene Symbol: Bhlhe40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011498.4 | NP_035628.1 | 2 - 3 | 438 | 121 | |
| GenBank mRNA | AF010305.1 | - | 2 - 3 | 370 | 121 | |
| AF364051.1 | - | 2 - 3 | 370 | 121 | ||
| AK048242.1 | - | 2 - 3 | 439 | 121 | ||
| AK090032.1 | - | 2 - 3 | 392 | 121 | ||
| AK154935.1 | - | 2 - 3 | 391 | 121 | ||
| BC010720.1 | - | 2 - 3 | 381 | 3707474 | 121 | |
| Y07836.1 | - | 2 - 3 | 133 | 121 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Bhlhe40
Genomic Map