Assay Details
| Gene Symbol: Pold2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008894.2 | NP_032920.2 | 3 - 4 | 465 | 96 | |
| GenBank mRNA | AK013901.1 | - | 3 - 4 | 432 | 96 | |
| AK218431.1 | - | 2 - 3 | 180 | 96 | ||
| BC003342.1 | - | 3 - 4 | 431 | 3257610 | 96 | |
| BG078511.2 | - | 3 - 4 | 418 | 96 | ||
| Z72486.1 | - | 3 - 4 | 426 | 96 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pold2
Genomic Map