Assay Details
| Gene Symbol: Pold2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008894.2 | NP_032920.2 | 4 - 5 | 588 | 69 | |
| GenBank mRNA | AK013901.1 | - | 4 - 5 | 555 | 69 | |
| AK218431.1 | - | 3 - 4 | 303 | 69 | ||
| BC003342.1 | - | 4 - 5 | 554 | 3257610 | 69 | |
| BG078511.2 | - | 4 - 5 | 541 | 69 | ||
| CB205181.1 | - | 1 - 2 | 124 | 69 | ||
| Z72486.1 | - | 4 - 5 | 549 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pold2
Genomic Map