Assay Details
| Gene Symbol: Gck | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001287386.1 | NP_001274315.1 | 1 - 2 | 171 | 58 | |
| GenBank mRNA | AK142698.1 | - | 1 - 2 | 170 | 58 | |
| BC011139.1 | - | 1 - 2 | 160 | 4195943 | 58 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gck
Genomic Map