Assay Details
| Gene Symbol: Dyrk4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_207210.2 | NP_997093.2 | 1 - 2 | 87 | 70 | |
| GenBank mRNA | AK077117.1 | - | 1 - 2 | 88 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dyrk4
Genomic Map