Assay Details
| Gene Symbol: Dyrk4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_207210.2 | NP_997093.2 | 3 - 4 | 258 | 65 | |
| GenBank mRNA | AK077117.1 | - | 3 - 4 | 259 | 65 | |
| BC052324.1 | - | 2 - 3 | 312 | 6315342 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dyrk4
Genomic Map