Assay Details
| Gene Symbol: Fgf6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010204.1 | NP_034334.1 | 1 - 2 | 389 | 66 | |
| GenBank mRNA | AK086530.1 | - | 1 - 2 | 307 | 66 | |
| M92416.1 | - | 1 - 2 | 291 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fgf6
Genomic Map