Assay Details
| Gene Symbol: Rad51ap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009013.3 | NP_033039.2 | 6 - 7 | 610 | 153 | |
| GenBank mRNA | AK011379.1 | - | 6 - 7 | 611 | 153 | |
| AK077829.1 | - | 6 - 7 | 640 | 153 | ||
| AK079525.1 | - | 6 - 7 | 632 | 153 | ||
| BC003738.1 | - | 6 - 7 | 624 | 3495632 | 153 | |
| U93583.1 | - | 6 - 7 | 608 | 153 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rad51ap1
Genomic Map