Assay Details
| Gene Symbol: Il17ra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008359.2 | NP_032385.1 | 2 - 3 | 297 | 120 | |
| GenBank mRNA | AK036587.1 | - | 2 - 3 | 249 | 120 | |
| AK080226.2 | - | 2 - 3 | 248 | 120 | ||
| U31993.1 | - | 2 - 3 | 285 | 120 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Il17ra
Genomic Map