Assay Details
| Gene Symbol: Renbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164704.1 | NP_001158176.1 | 8 - 9 | 1014 | 58 | |
| NM_023132.3 | NP_075621.3 | 8 - 9 | 1056 | 58 | ||
| GenBank mRNA | AK014347.1 | - | 8 - 9 | 976 | 58 | |
| AK146652.1 | - | 8 - 9 | 1021 | 58 | ||
| AK167648.1 | - | 8 - 9 | 927 | 58 | ||
| AY183138.1 | - | 8 - 9 | 1024 | 58 | ||
| BC014821.1 | - | 8 - 9 | 950 | 4238244 | 58 | |
| BC034846.1 | - | 4 - 5 | 1740 | 58 | ||
| BC035455.1 | - | 8 - 9 | 950 | 58 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Renbp
Genomic Map