Assay Details
| Gene Symbol: Taz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001173547.2 | NP_001167018.1 | 3 - 4 | 570 | 69 | |
| NM_001242615.2 | NP_001229544.1 | 3 - 4 | 570 | 69 | ||
| NM_001242616.2 | NP_001229545.1 | 3 - 4 | 570 | 69 | ||
| NM_001290738.1 | NP_001277667.1 | 3 - 4 | 570 | 69 | ||
| NM_181516.6 | NP_852657.1 | 3 - 4 | 570 | 69 | ||
| GenBank mRNA | AK004035.1 | - | 3 - 4 | 527 | 69 | |
| AK075615.1 | - | 3 - 4 | 527 | 69 | ||
| AK076481.1 | - | 3 - 4 | 538 | 69 | ||
| AK160327.1 | - | 3 - 4 | 539 | 69 | ||
| AK170662.1 | - | 3 - 4 | 539 | 69 | ||
| AK188231.1 | - | 1 - 2 | 28 | 69 | ||
| AY231459.1 | - | 3 - 4 | 289 | 69 | ||
| AY231460.1 | - | 3 - 4 | 289 | 69 | ||
| AY258040.1 | - | 3 - 4 | 289 | 69 | ||
| AY258041.1 | - | 3 - 4 | 289 | 69 | ||
| BC015305.1 | - | 3 - 4 | 535 | 4221041 | 69 | |
| CD807936.1 | - | 3 - 4 | 516 | 69 | ||
| EF524069.1 | - | 2 - 3 | 169 | 69 | ||
More Information
Set Membership: |
|

Taz
Genomic Map