Assay Details
| Gene Symbol: Klra1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF074456.1 | - | 3 - 4 | 403 | 92 | |
| AF218077.1 | - | 2 - 3 | 256 | 92 | ||
| AF374002.1 | - | 3 - 4 | 291 | 92 | ||
| AF374003.1 | - | 3 - 4 | 291 | 92 | ||
| U34890.1 | - | 2 - 3 | 209 | 92 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2004551 | BC144929.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2004551 | M25775.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2004551 | M25812.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2004551 | AK080158.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2004551 | AK172530.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 471582 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 293522 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 6664 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 19855 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 139984 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 18305 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P108063 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2004551 | BC144929.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2004551 | M25775.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2004551 | M25812.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2004551 | AK080158.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2004551 | AK172530.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06069919 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06069940 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06069959 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06069930 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05605777 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24847850 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24847842 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24847839 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24847834 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24847896 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24847844 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24847832 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24847853 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24847835 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24847866 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24847852 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24847820 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24847777 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24847782 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24847855 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24847846 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24847818 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24847861 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24847914 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24847911 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24847838 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05529797 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24847903 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24847910 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05382894 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24847896 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10548485 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10548494 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10548483 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10548458 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10548488 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10548493 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10548487 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10548490 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10548492 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10548484 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10548486 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10548495 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10548489 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1428056_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10548455 | - | - |
| Mouse Genome 430 2.0 Array | 1452645_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1452645_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1428056_at | - | - |
| Mouse Genome 430 2.0 Array | 1428056_PM_at | - | - |
| Mouse Expression Set 430 | 1452645_PM_x_at | - | - |
| Mouse Expression Set 430 | 1428056_PM_at | - | - |
| Mouse Expression Set 430 | 1452645_x_at | - | - |
| Mouse Expression Set 430 | 1428056_at | - | - |
| Murine Genome U74v2 Set | 95908_PM_at | - | - |
| Murine Genome U74v2 Set | 95908_at | - | - |
| Mouse Genome 430A 2.0 Array | 1428056_at | - | - |
| Mouse Genome 430A 2.0 Array | 1452645_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1428056_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1452645_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1428056_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1428056_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 106040338 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2370008 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 106040338 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002370008 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1233407 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1228875 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1233407 | - | - |
| mouse-6 v1.1 (Illumina) | GI_7710047-S | - | - |
| mouse-6 v1.1 (Illumina) | scl28320.9.1_99-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 106040338 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2370008 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl28320.9.1_99-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 106040338 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2370008 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1228875 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1228875 | - | - |

Klra1
Genomic Map