Assay Details
| Gene Symbol: Clec7a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020008.2 | NP_064392.2 | 3 - 4 | 427 | 83 | |
| GenBank mRNA | AF262985.1 | - | 3 - 4 | 423 | 83 | |
| AY534909.1 | - | 3 - 4 | 335 | 83 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Clec7a
Genomic Map