Assay Details
| Gene Symbol: Klre1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153590.3 | NP_705818.3 | 1 - 2 | 77 | 69 | |
| GenBank mRNA | AF306663.1 | - | 1 - 2 | 77 | 69 | |
| AY100458.1 | - | 1 - 2 | 470 | 69 | ||
| BC145177.1 | - | 1 - 2 | 112 | 69 | ||
| BG145575.1 | - | 1 - 2 | 55 | 69 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AF306663.1, AY100458.1, NM_153590.3 transcripts.
Set Membership: |
|

Klre1
Genomic Map