Assay Details
| Gene Symbol: Klre1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153590.3 | NP_705818.3 | 3 - 4 | 82 | ||
| GenBank mRNA | AF306663.1 | - | 3 - 4 | 82 | ||
| AY100458.1 | - | 3 - 4 | 773 | 82 | ||
| BC145177.1 | - | 3 - 4 | 82 | |||
More Information
Set Membership: |
|

Klre1
Genomic Map