Assay Details
| Gene Symbol: Slc6a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178703.4 | NP_848818.1 | 15 - 16 | 2074 | 84 | |
| GenBank mRNA | AK042956.1 | - | 15 - 16 | 1936 | 84 | |
| AK052971.1 | - | 15 - 16 | 1899 | 84 | ||
| AK053883.1 | - | 15 - 16 | 1896 | 84 | ||
| BC027111.1 | - | 2 - 3 | 269 | 84 | ||
| BC059080.1 | - | 15 - 16 | 1925 | 6827121 | 84 | |
| M92378.1 | - | 14 - 15 | 1833 | 84 | ||
More Information
Set Membership: |
|

Slc6a1
Genomic Map