Assay Details
| Gene Symbol: Tamm41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026894.1 | NP_081170.1 | 5 - 6 | 798 | 76 | |
| GenBank mRNA | AK005100.1 | - | 5 - 6 | 798 | 76 | |
| BE956902.1 | - | 3 - 4 | 76 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Tamm41
Genomic Map