Assay Details
| Gene Symbol: Mxd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010751.3 | NP_034881.2 | 2 - 3 | 316 | 69 | |
| GenBank mRNA | AK151014.1 | - | 2 - 3 | 278 | 69 | |
| AK151114.1 | - | 2 - 3 | 277 | 69 | ||
| AK151305.1 | - | 2 - 3 | 315 | 69 | ||
| AK151882.1 | - | 2 - 3 | 277 | 69 | ||
| AK153303.1 | - | 2 - 3 | 277 | 69 | ||
| BC034860.1 | - | 2 - 3 | 253 | 1448816 | 69 | |
| CT010233.1 | - | 2 - 3 | 165 | 69 | ||
| L38926.1 | - | 2 - 3 | 201 | 69 | ||
| U20614.1 | - | 2 - 3 | 270 | 69 | ||
| X83106.1 | - | 2 - 3 | 214 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mxd1
Genomic Map