Assay Details
| Gene Symbol: Gmcl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011818.3 | NP_035948.3 | 11 - 12 | 1426 | 82 | |
| GenBank mRNA | AF163665.1 | - | 11 - 12 | 1370 | 82 | |
| AF186095.1 | - | 11 - 12 | 1262 | 82 | ||
| AF282322.1 | - | 11 - 12 | 1296 | 82 | ||
| AK004716.1 | - | 11 - 12 | 1402 | 82 | ||
| AK146964.1 | - | 11 - 12 | 1395 | 82 | ||
| AK155238.1 | - | 11 - 12 | 1409 | 82 | ||
| BC054767.1 | - | 11 - 12 | 1385 | 6414943 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gmcl1
Genomic Map