Assay Details
| Gene Symbol: Mgll | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166249.1 | NP_001159721.1 | 4 - 5 | 548 | 65 | |
| NM_001166251.1 | NP_001159723.1 | 4 - 5 | 863 | 65 | ||
| NM_011844.4 | NP_035974.1 | 4 - 5 | 548 | 65 | ||
| GenBank mRNA | AJ001118.2 | - | 4 - 5 | 476 | 65 | |
| AJ316580.1 | - | 4 - 5 | 739 | 65 | ||
| AK131645.1 | - | 4 - 5 | 528 | 65 | ||
| AK148242.1 | - | 4 - 5 | 491 | 65 | ||
| AK148395.1 | - | 4 - 5 | 491 | 65 | ||
| BC057965.1 | - | 4 - 5 | 473 | 4986023 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mgll
Genomic Map