Assay Details
| Gene Symbol: Eefsec | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023060.3 | NP_075547.1 | 2 - 3 | 545 | 85 | |
| GenBank mRNA | AF268871.1 | - | 2 - 3 | 509 | 85 | |
| AK028424.1 | - | 2 - 3 | 527 | 85 | ||
| AK087501.1 | - | 2 - 3 | 533 | 85 | ||
| AK147924.1 | - | 2 - 3 | 514 | 85 | ||
| AK171885.1 | - | 2 - 3 | 525 | 85 | ||
| BC024915.1 | - | 2 - 3 | 519 | 3158026 | 85 | |
| BC099675.1 | - | 2 - 3 | 503 | 5011896 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

Eefsec
Genomic Map