Assay Details
| Gene Symbol: Eefsec | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023060.3 | NP_075547.1 | 3 - 4 | 643 | 61 | |
| GenBank mRNA | AF268871.1 | - | 3 - 4 | 607 | 61 | |
| AF283518.1 | - | 3 - 4 | 598 | 61 | ||
| AK028424.1 | - | 3 - 4 | 625 | 61 | ||
| AK087501.1 | - | 3 - 4 | 631 | 61 | ||
| AK147924.1 | - | 3 - 4 | 612 | 61 | ||
| AK171885.1 | - | 3 - 4 | 623 | 61 | ||
| AK184750.1 | - | 1 - 2 | 97 | 61 | ||
| AK207457.1 | - | 2 - 3 | 311 | 61 | ||
| BC024915.1 | - | 3 - 4 | 617 | 3158026 | 61 | |
| BC099675.1 | - | 3 - 4 | 601 | 5011896 | 61 | |
More Information
Updated Mapping Information
Set Membership: |
|

Eefsec
Genomic Map