Assay Details
| Gene Symbol: Eefsec | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023060.3 | NP_075547.1 | 4 - 5 | 808 | 97 | |
| GenBank mRNA | AF268871.1 | - | 4 - 5 | 772 | 97 | |
| AF283518.1 | - | 4 - 5 | 763 | 97 | ||
| AK028424.1 | - | 4 - 5 | 790 | 97 | ||
| AK087501.1 | - | 4 - 5 | 796 | 97 | ||
| AK147924.1 | - | 4 - 5 | 777 | 97 | ||
| AK171885.1 | - | 4 - 5 | 788 | 97 | ||
| AK184750.1 | - | 2 - 3 | 262 | 97 | ||
| BC024915.1 | - | 4 - 5 | 782 | 3158026 | 97 | |
| BC099675.1 | - | 4 - 5 | 766 | 5011896 | 97 | |
More Information
Updated Mapping Information
Set Membership: |
|

Eefsec
Genomic Map