Assay Details
| Gene Symbol: Antxr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_054041.2 | NP_473382.1 | 6 - 7 | 817 | 71 | |
| GenBank mRNA | AF378762.1 | - | 6 - 7 | 757 | 71 | |
| AK013005.1 | - | 4 - 5 | 224 | 71 | ||
| AK031465.1 | - | 6 - 7 | 807 | 71 | ||
| AK048637.1 | - | 6 - 7 | 864 | 71 | ||
| BC057043.1 | - | 4 - 5 | 255 | 71 | ||
| BC094544.1 | - | 6 - 7 | 762 | 6406827 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Antxr1
Genomic Map