Assay Details
| Gene Symbol: Gkn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026860.1 | NP_081136.1 | 2 - 3 | 191 | 88 | |
| GenBank mRNA | AK004474.1 | - | 2 - 3 | 111 | 88 | |
| BC116955.1 | - | 2 - 3 | 191 | 40126274 | 88 | |
| FN600638.1 | - | 2 - 3 | 110 | 88 | ||
| GU220566.1 | - | 2 - 3 | 110 | 88 | ||
More Information
Set Membership: |
|

Gkn3
Genomic Map