Assay Details
| Gene Symbol: Gkn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026860.1 | NP_081136.1 | 4 - 5 | 440 | 75 | |
| GenBank mRNA | AK004474.1 | - | 4 - 5 | 360 | 75 | |
| BC116955.1 | - | 4 - 5 | 440 | 40126274 | 75 | |
| FN600638.1 | - | 4 - 5 | 359 | 75 | ||
| GU220566.1 | - | 4 - 5 | 359 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gkn3
Genomic Map