Assay Details
| Gene Symbol: Gkn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026860.1 | NP_081136.1 | 5 - 6 | 585 | 67 | |
| GenBank mRNA | AK004474.1 | - | 5 - 6 | 505 | 67 | |
| BC116955.1 | - | 5 - 6 | 585 | 40126274 | 67 | |
| FN600638.1 | - | 5 - 6 | 504 | 67 | ||
| GU220566.1 | - | 5 - 6 | 504 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P267700 | GU220566.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P267700 | NM_026860.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P267700 | AK004474.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P267700 | BC116955.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P267700 | FN600638.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P267700 | GU220566.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P267700 | NM_026860.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P267700 | AK004474.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P267700 | BC116955.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P267700 | FN600638.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 724113 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 753319 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P123397 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P267700 | GU220566.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P267700 | NM_026860.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P267700 | AK004474.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P267700 | BC116955.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P267700 | FN600638.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P267700 | GU220566.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P267700 | NM_026860.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P267700 | AK004474.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P267700 | BC116955.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P267700 | FN600638.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05971394 | - | - |
| Whole Mouse Genome G4122A | A_51_P267700 | GU220566.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P267700 | NM_026860.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P267700 | AK004474.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P267700 | BC116955.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P267700 | FN600638.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P267700 | GU220566.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P267700 | NM_026860.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P267700 | AK004474.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P267700 | BC116955.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P267700 | FN600638.1 | mouse_transcriptome |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24615736 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24615816 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24615827 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24615695 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24615778 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24615738 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24615811 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24615751 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24615833 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24615784 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24615814 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24615709 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24615653 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24615813 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24615820 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24615775 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24615684 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24615671 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24615728 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24615817 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24615725 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24615802 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24615707 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24615781 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24615830 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24615763 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24615822 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24615681 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24615766 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24615804 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24615732 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24615758 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24615794 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24615674 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24615666 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24615795 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24615788 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24615660 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24615689 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24615790 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24615694 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24615696 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24615770 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24615678 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24615764 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24615692 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24615753 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24615669 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24615723 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24615759 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24615771 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24615734 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24615657 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24615777 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24615762 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24615679 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24615724 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24615803 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24615655 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24615799 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24615773 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24615673 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24615690 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24615727 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24615797 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24615718 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24615683 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24615786 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24615664 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24615740 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24615705 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24615712 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24615828 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24615662 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24615779 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24615719 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24615697 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24615676 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24615835 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24615703 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24615768 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24615749 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24615782 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24615711 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24615700 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24615721 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24615652 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24615701 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24615757 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24615687 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24615788 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24615778 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24615773 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05813208 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24615763 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05542024 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05631397 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24615736 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05128402 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24615811 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24615771 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24615814 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24615756 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24615784 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24615795 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24615802 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24615765 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05382742 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24615797 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24615751 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24615770 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24615739 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24615724 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24615790 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24615734 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24615749 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24615753 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24615759 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24615764 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24615799 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24615782 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05705887 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24615781 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24615807 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24615747 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24615779 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24615801 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24615725 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24615786 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24615757 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05435430 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24615728 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24615752 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24615718 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24615731 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24615804 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24615732 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05510343 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24615758 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05510341 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05382751 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24615815 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24615808 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24615719 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24615755 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24615740 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24615793 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24615723 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24615794 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05631398 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05115911 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24615727 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10546005 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10546002 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10546009 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10546007 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10546008 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10546003 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10546004 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10546006 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1438648_PM_x_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1429286_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10546001 | - | - |
| Mouse Genome 430 2.0 Array | 1438648_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1438648_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1429286_at | - | - |
| Mouse Genome 430 2.0 Array | 1429286_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 68888_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 68888_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0600020628.mm.1 | TC0600002654.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600020628.mm.1 | NM_026860.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600020628.mm.1 | NM_026860.1 | Refseq |
| Mouse Expression Set 430 | 1429286_at | - | - |
| Mouse Expression Set 430 | 1438648_PM_x_at | - | - |
| Mouse Expression Set 430 | 1438648_x_at | - | - |
| Mouse Expression Set 430 | 1429286_PM_at | - | - |
| Murine Genome U74v2 Set | 165955_PM_r_at | - | - |
| Murine Genome U74v2 Set | 165811_PM_at | - | - |
| Murine Genome U74v2 Set | 165955_r_at | - | - |
| Murine Genome U74v2 Set | 165811_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1438648_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1438648_x_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1429286_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1429286_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 861759 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseRef-8 v2 Expression BeadChip | ILMN_2856588 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2856588 | - | - |

Gkn3
Genomic Map