Assay Details
| Gene Symbol: Rad52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166381.1 | NP_001159853.1 | 8 - 9 | 894 | 85 | |
| NM_001166382.1 | NP_001159854.1 | 8 - 9 | 894 | 85 | ||
| NM_001166383.1 | NP_001159855.1 | 8 - 9 | 894 | 85 | ||
| NM_011236.2 | NP_035366.2 | 8 - 9 | 894 | 85 | ||
| GenBank mRNA | AK080702.1 | - | 8 - 9 | 865 | 85 | |
| BC010673.1 | - | 7 - 8 | 766 | 85 | ||
| BC019123.1 | - | 8 - 9 | 874 | 5003696 | 85 | |
| U06837.1 | - | 8 - 9 | 816 | 85 | ||
| U12135.1 | - | 8 - 9 | 863 | 85 | ||
| Z32767.1 | - | 8 - 9 | 888 | 85 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rad52
Genomic Map