Assay Details
| Gene Symbol: Erc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053204.2 | NP_444434.2 | 18 - 19 | 3505 | 70 | |
| NR_104484.1 | 17 - 18 | 3373 | 70 | |||
| GenBank mRNA | AF340028.1 | - | 17 - 18 | 3357 | 70 | |
| AF340029.1 | - | 18 - 19 | 3489 | 70 | ||
| AK122437.1 | - | 6 - 7 | 736 | 70 | ||
| AY316692.1 | - | 16 - 17 | 2937 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Erc1
Genomic Map