Assay Details
| Gene Symbol: Adipor2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_197985.3 | NP_932102.2 | 2 - 3 | 326 | 90 | |
| GenBank mRNA | AK039667.1 | - | 2 - 3 | 534 | 90 | |
| AK046591.1 | - | 2 - 3 | 333 | 90 | ||
| AK134787.1 | - | 2 - 3 | 342 | 90 | ||
| AK148295.1 | - | 2 - 3 | 468 | 90 | ||
| AK160534.1 | - | 2 - 3 | 343 | 90 | ||
| AK161404.1 | - | 2 - 3 | 291 | 90 | ||
| AY424291.1 | - | 2 - 3 | 326 | 90 | ||
| BC024094.1 | - | 2 - 3 | 326 | 5132696 | 90 | |
| BC064109.1 | - | 2 - 3 | 324 | 3371697 | 90 | |
More Information
Updated Mapping Information
Set Membership: |
|

Adipor2
Genomic Map