Assay Details
| Gene Symbol: Adipor2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_197985.3 | NP_932102.2 | 5 - 6 | 797 | 79 | |
| GenBank mRNA | AK039667.1 | - | 5 - 6 | 1005 | 79 | |
| AK046591.1 | - | 5 - 6 | 804 | 79 | ||
| AK134787.1 | - | 5 - 6 | 813 | 79 | ||
| AK148295.1 | - | 5 - 6 | 939 | 79 | ||
| AK160534.1 | - | 5 - 6 | 814 | 79 | ||
| AK161404.1 | - | 6 - 7 | 822 | 79 | ||
| AY424291.1 | - | 5 - 6 | 797 | 79 | ||
| BC024094.1 | - | 5 - 6 | 797 | 5132696 | 79 | |
| BC064109.1 | - | 5 - 6 | 795 | 3371697 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Adipor2
Genomic Map