Assay Details
| Gene Symbol: Klrg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016970.1 | NP_058666.1 | 2 - 3 | 206 | 86 | |
| GenBank mRNA | AF097357.1 | - | 2 - 3 | 206 | 86 | |
| AJ010751.1 | - | 2 - 3 | 255 | 86 | ||
| BC101953.1 | - | 2 - 3 | 205 | 40044827 | 86 | |
| BC103546.1 | - | 2 - 3 | 205 | 40044831 | 86 | |
| BC103547.1 | - | 2 - 3 | 205 | 40044830 | 86 | |
| BC103548.1 | - | 2 - 3 | 205 | 40044829 | 86 | |
More Information
Updated Mapping Information
Set Membership: |
|

Klrg1
Genomic Map