Assay Details
| Gene Symbol: Phc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042623.2 | NP_001036088.1 | 6 - 7 | 1052 | 108 | |
| NM_001271579.1 | NP_001258508.1 | 7 - 8 | 1179 | 108 | ||
| NM_007905.3 | NP_031931.2 | 7 - 8 | 1208 | 108 | ||
| NR_073364.1 | 7 - 8 | 1215 | 108 | |||
| GenBank mRNA | AK036370.1 | - | 7 - 8 | 1168 | 108 | |
| AK132756.1 | - | 6 - 7 | 1010 | 108 | ||
| AK167590.1 | - | 6 - 7 | 1041 | 108 | ||
| BC046535.1 | - | 7 - 8 | 1189 | 4935713 | 108 | |
| BC052394.1 | - | 7 - 8 | 1541 | 5719837 | 108 | |
| S73882.1 | - | 7 - 8 | 1164 | 108 | ||
| S73883.1 | - | 7 - 8 | 1171 | 108 | ||
| S73884.1 | - | 7 - 8 | 1171 | 108 | ||
| U63386.1 | - | 6 - 7 | 1130 | 108 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Phc1
Genomic Map