Assay Details
| Gene Symbol: Pex26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK014598.1 | - | 2 - 3 | 739 | 65 | |
| AK040411.1 | - | 729 | 65 | |||
| AK042663.1 | - | 2 - 3 | 739 | 65 | ||
| AK044186.1 | - | 2 - 3 | 691 | 65 | ||
| AK087637.1 | - | 2 - 3 | 961 | 65 | ||
| AK147925.1 | - | 2 - 3 | 731 | 65 | ||
| BC019144.1 | - | 2 - 3 | 722 | 5050939 | 65 | |
| BC058694.1 | - | 2 - 3 | 712 | 6811976 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pex26
Genomic Map