Assay Details
| Gene Symbol: Pparg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011146.3 | NP_035276.2 | 5 - 6 | 858 | 139 | |
| GenBank mRNA | AB644275.1 | - | 6 - 7 | 864 | 139 | |
| AY208183.1 | - | 5 - 6 | 858 | 139 | ||
| AY208184.1 | - | 5 - 6 | 858 | 139 | ||
| AY243585.1 | - | 5 - 6 | 858 | 139 | ||
| BC021798.1 | - | 5 - 6 | 858 | 4191539 | 139 | |
| CT010340.1 | - | 5 - 6 | 813 | 139 | ||
| GQ868647.1 | - | 4 - 5 | 616 | 139 | ||
| U01664.1 | - | 6 - 7 | 890 | 139 | ||
| U01841.1 | - | 6 - 7 | 1213 | 139 | ||
| U09138.1 | - | 5 - 6 | 852 | 139 | ||
| U10374.1 | - | 4 - 5 | 723 | 139 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Citations (1)
Increased adiposity in the retinol saturase-knockout mouse.
- Authors
- Moise AR, Lobo GP, Erokwu B, Wilson DL, Peck D, Alvarez S, Dominguez M, Alvarez R, Flask CA, de Lera AR, von Lintig J, Palczewski K
- Journal
- Faseb J - Page No 1261 (Thu Apr 01 00:00:00 EDT 2010)
- Volume (Year)
- 24 (2010)
- Product Usage
- -

Pparg
Genomic Map