Assay Details
| Gene Symbol: Syn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111015.1 | NP_001104485.1 | 9 - 10 | 1346 | 82 | |
| NM_013681.3 | NP_038709.1 | 9 - 10 | 1346 | 82 | ||
| GenBank mRNA | AF096867.1 | - | 9 - 10 | 1347 | 82 | |
| AK029181.1 | - | 7 - 8 | 688 | 82 | ||
| AK043584.1 | - | 4 - 5 | 1558 | 82 | ||
| BC053427.1 | - | 9 - 10 | 1267 | 82 | ||
| BC066004.1 | - | 9 - 10 | 1165 | 82 | ||
| BC085129.1 | - | 9 - 10 | 1165 | 5701861 | 82 | |
| BM951908.1 | - | 3 - 4 | 220 | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Syn2
Genomic Map