Assay Details
| Gene Symbol: Raf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029780.3 | NP_084056.1 | 7 - 8 | 1256 | 65 | |
| GenBank mRNA | AB057655.1 | - | 6 - 7 | 801 | 65 | |
| AB057663.2 | - | 7 - 8 | 1026 | 65 | ||
| AK141745.1 | - | 7 - 8 | 1257 | 65 | ||
| AK215463.1 | - | 1 - 2 | 58 | 65 | ||
| BC015273.1 | - | 7 - 8 | 1135 | 4222659 | 65 | |
| BC056439.1 | - | 7 - 8 | 1175 | 65 | ||
| BC092040.1 | - | 7 - 8 | 1121 | 2645313 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Raf1
Genomic Map