Assay Details
| Gene Symbol: Foxm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008021.4 | NP_032047.4 | 1 - 2 | 123 | 66 | |
| GenBank mRNA | AK041638.1 | - | 1 - 2 | 91 | 66 | |
| AK049177.1 | - | 1 - 2 | 64 | 66 | ||
| BC065067.1 | - | 1 - 2 | 123 | 6417437 | 66 | |
| BC075723.1 | - | 1 - 2 | 142 | 6515008 | 66 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK049177.1, BC065067.1, BC075723.1, NM_008021.4 transcripts.
Set Membership: |
|

Foxm1
Genomic Map