Assay Details
| Gene Symbol: Foxm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008021.4 | NP_032047.4 | 6 - 7 | 1188 | 66 | |
| GenBank mRNA | AK049177.1 | - | 6 - 7 | 1129 | 66 | |
| AK213171.1 | - | 3 - 4 | 334 | 66 | ||
| BC065067.1 | - | 6 - 7 | 1188 | 6417437 | 66 | |
| BC075723.1 | - | 6 - 7 | 1207 | 6515008 | 66 | |
| Y11245.1 | - | 6 - 7 | 1096 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Foxm1
Genomic Map