Assay Details
| Gene Symbol: Fkbp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010219.3 | NP_034349.1 | 6 - 7 | 881 | 66 | |
| GenBank mRNA | AK083912.1 | - | 6 - 7 | 881 | 66 | |
| AK089972.1 | - | 6 - 7 | 890 | 66 | ||
| AK159211.1 | - | 6 - 7 | 908 | 66 | ||
| AK160202.1 | - | 6 - 7 | 830 | 66 | ||
| BC003447.1 | - | 6 - 7 | 898 | 2651490 | 66 | |
| X70887.1 | - | 6 - 7 | 778 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fkbp4
Genomic Map