Assay Details
| Gene Symbol: Clec2i | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289707.1 | NP_001276636.1 | 1 - 2 | 78 | 70 | |
| NM_020257.2 | NP_064653.1 | 1 - 2 | 78 | 70 | ||
| GenBank mRNA | AF121352.1 | - | 1 - 2 | 51 | 70 | |
| AF350411.1 | - | 1 - 2 | 55 | 70 | ||
| AK041724.1 | - | 1 - 2 | 76 | 70 | ||
| AY137344.1 | - | 1 - 2 | 77 | 70 | ||
| AY137345.1 | - | 1 - 2 | 77 | 70 | ||
| AY256575.1 | - | 1 - 2 | 67 | 70 | ||
| BC139192.1 | - | 1 - 2 | 51 | 8862214 | 70 | |
| BC139193.1 | - | 1 - 2 | 51 | 8862215 | 70 | |
| BC145574.1 | - | 1 - 2 | 51 | 9054140 | 70 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AF350411.1 transcripts.
Set Membership: |
|

Clec2i
Genomic Map