Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P589119 |
AK168313.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P589119 |
BC027368.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P589119 |
BE655013.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P589119 |
AK146167.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P589119 |
AK156717.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P589119 |
AK168313.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P589119 |
BC027368.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P589119 |
BE655013.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P589119 |
AK146167.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P589119 |
AK156717.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
535471 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
186516 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
825411 |
AK035807.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
825411 |
NM_001110203.1 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
969810 |
- |
- |
Whole Mouse Genome G4122A |
A_52_P268549 |
AK168313.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P268549 |
BC005726.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P268549 |
BC027368.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P268549 |
AK146167.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P268549 |
AK156717.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P589119 |
AK168313.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P589119 |
BC027368.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P589119 |
BE655013.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P589119 |
AK146167.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P589119 |
AK156717.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P589119 |
AK168313.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P589119 |
BC027368.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P589119 |
BE655013.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P589119 |
AK146167.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P589119 |
AK156717.1 |
mouse_transcriptome |