Assay Details
| Gene Symbol: Itfg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133927.1 | NP_598688.1 | 4 - 5 | 503 | 64 | |
| GenBank mRNA | AK134630.1 | - | 4 - 5 | 524 | 64 | |
| AK147728.1 | - | 4 - 5 | 505 | 64 | ||
| AK159637.1 | - | 4 - 5 | 512 | 64 | ||
| BC014833.1 | - | 4 - 5 | 503 | 4507661 | 64 | |
| BC025492.1 | - | 4 - 5 | 523 | 5309950 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Itfg2
Genomic Map