Assay Details
| Gene Symbol: Atp2b2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036684.2 | NP_001031761.1 | 3 - 4 | 841 | 75 | |
| NM_009723.5 | NP_033853.1 | 5 - 6 | 1030 | 75 | ||
| GenBank mRNA | AB030737.1 | - | 2 - 3 | 402 | 75 | |
| AB030738.1 | - | 2 - 3 | 402 | 75 | ||
| AK051287.1 | - | 3 - 4 | 836 | 75 | ||
| AK054112.1 | - | 4 - 5 | 992 | 75 | ||
| AK147240.1 | - | 5 - 6 | 916 | 75 | ||
| AK147357.1 | - | 3 - 4 | 770 | 75 | ||
| AK147369.1 | - | 5 - 6 | 966 | 75 | ||
| AK147399.1 | - | 3 - 4 | 1005 | 75 | ||
| AK147501.1 | - | 5 - 6 | 977 | 75 | ||
| BC075643.1 | - | 3 - 4 | 841 | 30621322 | 75 | |
| GU734816.2 | - | 2 - 3 | 402 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Atp2b2
Genomic Map